siRNA Details
| Gene Symbol: Kcnq5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134643.2 | NP_001128115.2 | 5 | 1056 | ||
| GenBank mRNA | FQ084615.1 | - | 5 | 697 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnq5| Gene Symbol: Kcnq5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134643.2 | NP_001128115.2 | 5 | 1056 | ||
| GenBank mRNA | FQ084615.1 | - | 5 | 697 | ||