siRNA Details
| Gene Symbol: Brs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152845.1 | NP_690058.1 | 1 | 211 | ||
| XM_006257677.2 | XP_006257739.1 | 2 | 241 | |||
| XM_006257678.2 | XP_006257740.1 | 2 | 60 | |||
| GenBank mRNA | AF510984.1 | - | 1 | 211 | ||
More Information
Set Membership: |
|

Brs3
Genomic Map