siRNA Details
| Gene Symbol: Ly49i2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152848.1 | NP_690061.1 | 4 | 418 | ||
| GenBank mRNA | AY115572.1 | - | 4 | 418 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ly49i2| Gene Symbol: Ly49i2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152848.1 | NP_690061.1 | 4 | 418 | ||
| GenBank mRNA | AY115572.1 | - | 4 | 418 | ||