siRNA Details
| Gene Symbol: Aurka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153296.2 | NP_695208.2 | 7, 8 | 1018 | ||
| XM_006235675.2 | XP_006235737.1 | 8, 9 | 1097 | |||
| XM_006235676.2 | XP_006235738.1 | 8, 9 | 1114 | |||
| GenBank mRNA | AF537333.1 | - | 7, 8 | 1006 | ||
| BC104676.1 | - | 7, 8 | 1018 | 7453115 | ||
| FQ227101.1 | - | 8, 9 | 1103 | |||

Aurka
Genomic Map