siRNA Details
| Gene Symbol: Clrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153299.2 | NP_695211.2 | 3 | 583 | ||
| GenBank mRNA | AF482698.1 | - | 3 | 583 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clrn1| Gene Symbol: Clrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153299.2 | NP_695211.2 | 3 | 583 | ||
| GenBank mRNA | AF482698.1 | - | 3 | 583 | ||