siRNA Details
Gene Symbol: Csdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170542.1 | NP_001164013.1 | 4 | 661 | ||
GenBank mRNA | DY318921.1 | - | 2 | 61 | ||
X89962.1 | - | 4 | 560 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Csdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170542.1 | NP_001164013.1 | 4 | 661 | ||
GenBank mRNA | DY318921.1 | - | 2 | 61 | ||
X89962.1 | - | 4 | 560 |