siRNA Details
| Gene Symbol: Tomm20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152935.1 | NP_690918.1 | 3 | 317 | ||
| GenBank mRNA | BC078715.1 | - | 3 | 293 | 7127457 | |
| D63411.1 | - | 3 | 306 | |||
| FQ224957.1 | - | 3 | 334 | |||
| FQ230650.1 | - | 3 | 333 | |||
| U21871.1 | - | 3 | 317 | |||

Tomm20
Genomic Map