siRNA Details
| Gene Symbol: Aldh1a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153300.1 | NP_695212.1 | 11, 12 | 1400 | ||
| GenBank mRNA | AB369261.1 | - | 11, 12 | 1400 | ||
| AF434845.1 | - | 11, 12 | 1400 | |||
| BC166415.1 | - | 1457 | 9025597 | |||

Aldh1a3
Genomic Map