siRNA Details
| Gene Symbol: Tor1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153303.2 | NP_695215.2 | 2 | 384 | ||
| GenBank mRNA | AY054303.1 | - | 2 | 366 | ||
| BC078714.1 | - | 2 | 384 | 7134604 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tor1a| Gene Symbol: Tor1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153303.2 | NP_695215.2 | 2 | 384 | ||
| GenBank mRNA | AY054303.1 | - | 2 | 366 | ||
| BC078714.1 | - | 2 | 384 | 7134604 | ||