siRNA Details
| Gene Symbol: Ffar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153304.2 | NP_695216.1 | 1 | 570 | ||
| GenBank mRNA | AB095744.1 | - | 1 | 568 | ||
| AF539810.1 | - | 1 | 572 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ffar1| Gene Symbol: Ffar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153304.2 | NP_695216.1 | 1 | 570 | ||
| GenBank mRNA | AB095744.1 | - | 1 | 568 | ||
| AF539810.1 | - | 1 | 572 | |||