siRNA Details
| Gene Symbol: Ptpn9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013040.1 | NP_001013058.1 | 11 | 1310 | ||
| XM_006243105.2 | XP_006243167.1 | 11 | 1887 | |||
| XM_006243106.2 | XP_006243168.1 | 11 | 1902 | |||
| GenBank mRNA | BC082041.1 | - | 11 | 1310 | 7131532 | |

Ptpn9
Genomic Map