siRNA Details
| Gene Symbol: Slc4a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152938.1 | NP_690921.1 | 19 | 2587 | ||
| XM_006254530.2 | XP_006254592.1 | 19 | 2731 | |||
| XM_006254533.2 | XP_006254595.1 | 18 | 2659 | |||
| XM_008772020.1 | XP_008770242.1 | 19 | 2731 | |||
| XM_008772021.1 | XP_008770243.1 | 19 | 2732 | |||
| XM_008772023.1 | XP_008770245.1 | 18 | 2517 | |||
| GenBank mRNA | AB024339.1 | - | 19 | 2587 | ||

Slc4a9
Genomic Map