siRNA Details
| Gene Symbol: Grina | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153308.4 | NP_695220.4 | 7 | 1374 | ||
| GenBank mRNA | BC062074.1 | - | 7 | 1344 | 5599428 | |
| S61973.1 | - | 7 | 1325 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grina| Gene Symbol: Grina | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153308.4 | NP_695220.4 | 7 | 1374 | ||
| GenBank mRNA | BC062074.1 | - | 7 | 1344 | 5599428 | |
| S61973.1 | - | 7 | 1325 | |||