siRNA Details
Gene Symbol: Grina | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153308.4 | NP_695220.4 | 7 | 1168 | ||
GenBank mRNA | BC062074.1 | - | 7 | 1138 | 5599428 | |
S61973.1 | - | 7 | 1118 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Grina | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153308.4 | NP_695220.4 | 7 | 1168 | ||
GenBank mRNA | BC062074.1 | - | 7 | 1138 | 5599428 | |
S61973.1 | - | 7 | 1118 |