siRNA Details
Gene Symbol: Mina | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153309.2 | NP_695221.1 | 1024 | |||
GenBank mRNA | AB083195.1 | - | 999 | |||
BC087650.1 | - | 1024 | 7310399 |
Gene Symbol: LOC689439 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761010.1 | XP_008759232.1 | 1 | 1067 |
