siRNA Details
| Gene Symbol: Cyp4a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031605.2 | NP_113793.2 | 2 | 226 | ||
| GenBank mRNA | BC081771.1 | - | 2 | 226 | 7108258 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp4a8| Gene Symbol: Cyp4a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031605.2 | NP_113793.2 | 2 | 226 | ||
| GenBank mRNA | BC081771.1 | - | 2 | 226 | 7108258 | |