siRNA Details
Gene Symbol: Tmprss5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153311.1 | NP_695223.1 | 4, 5 | 403 | ||
GenBank mRNA | AF537098.1 | - | 4, 5 | 403 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tmprss5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153311.1 | NP_695223.1 | 4, 5 | 403 | ||
GenBank mRNA | AF537098.1 | - | 4, 5 | 403 |