siRNA Details
| Gene Symbol: Tmprss5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153311.1 | NP_695223.1 | 10 | 1074 | ||
| XM_008766194.1 | XP_008764416.1 | 8 | 937 | |||
| GenBank mRNA | AF537098.1 | - | 10 | 1074 | ||
| AF537099.1 | - | 8 | 852 | |||

Tmprss5
Genomic Map