siRNA Details
| Gene Symbol: Ugt2b17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153314.2 | NP_695226.2 | 908 | |||
| XM_006250800.2 | XP_006250862.1 | 2, 3 | 2002 | |||
| XM_008769998.1 | XP_008768220.1 | 2, 3 | 2008 | |||
| GenBank mRNA | M31109.1 | - | 908 | |||
| Y00156.1 | - | 2, 3 | 952 | |||

Ugt2b17
Genomic Map