siRNA Details
| Gene Symbol: Rnf40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153471.1 | NP_703201.1 | 15 | 2256 | ||
| XM_006230219.2 | XP_006230281.1 | 15 | 2334 | |||
| GenBank mRNA | AF352815.1 | - | 15 | 2256 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf40| Gene Symbol: Rnf40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153471.1 | NP_703201.1 | 15 | 2256 | ||
| XM_006230219.2 | XP_006230281.1 | 15 | 2334 | |||
| GenBank mRNA | AF352815.1 | - | 15 | 2256 | ||