siRNA Details
Gene Symbol: Mnat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153472.2 | NP_703202.1 | 5 | 543 | ||
GenBank mRNA | AY135567.1 | - | 5 | 498 | ||
BC078712.1 | - | 5 | 543 | 7128354 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mnat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153472.2 | NP_703202.1 | 5 | 543 | ||
GenBank mRNA | AY135567.1 | - | 5 | 498 | ||
BC078712.1 | - | 5 | 543 | 7128354 |