siRNA Details
| Gene Symbol: Fzd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153474.1 | NP_703204.1 | 4 | 919 | ||
| GenBank mRNA | AF323956.1 | - | 4 | 919 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fzd3| Gene Symbol: Fzd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153474.1 | NP_703204.1 | 4 | 919 | ||
| GenBank mRNA | AF323956.1 | - | 4 | 919 | ||
Set Membership: |
|