siRNA Details
| Gene Symbol: Dab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153621.1 | NP_705885.1 | 12 | 1517 | ||
| XM_008763854.1 | XP_008762076.1 | 15 | 2177 | |||
| GenBank mRNA | AB072426.1 | - | 12 | 1517 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dab1| Gene Symbol: Dab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153621.1 | NP_705885.1 | 12 | 1517 | ||
| XM_008763854.1 | XP_008762076.1 | 15 | 2177 | |||
| GenBank mRNA | AB072426.1 | - | 12 | 1517 | ||