siRNA Details
| Gene Symbol: Gpr64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270871.1 | NP_001257800.1 | 20, 21 | 1951 | ||
| NM_001270872.1 | NP_001257801.1 | 19, 20 | 1915 | |||
| NM_181366.2 | NP_852031.1 | 21, 22 | 1999 | |||
| XM_006256916.2 | XP_006256978.1 | 21, 22 | 2039 | |||
| XM_008773154.1 | XP_008771376.1 | 20, 21 | 2379 | |||
| XM_008773155.1 | XP_008771377.1 | 20, 21 | 1989 | |||
| XM_008773156.1 | XP_008771378.1 | 19, 20 | 2343 | |||
| XM_008773157.1 | XP_008771379.1 | 19, 20 | 2337 | |||
| XM_008773158.1 | XP_008771380.1 | 18, 19 | 2301 | |||
| XM_008773159.1 | XP_008771381.1 | 17, 18 | 1892 | |||
| XM_008773160.1 | XP_008771382.1 | 20, 21 | 2380 | |||
| GenBank mRNA | AF538953.1 | - | 21, 22 | 2009 | ||
| AF538958.1 | - | 20, 21 | 1961 | |||
| AF538959.1 | - | 19, 20 | 1925 | |||
More Information
Set Membership: |
|

Gpr64
Genomic Map