siRNA Details
| Gene Symbol: Nalcn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153630.1 | NP_705894.1 | 1956 | |||
| GenBank mRNA | AF078779.1 | - | 1956 | |||
| AY555273.1 | - | 1974 | ||||
| EF427668.1 | - | 15 | 1914 | |||
| EF492985.1 | - | 15 | 1914 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nalcn| Gene Symbol: Nalcn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153630.1 | NP_705894.1 | 1956 | |||
| GenBank mRNA | AF078779.1 | - | 1956 | |||
| AY555273.1 | - | 1974 | ||||
| EF427668.1 | - | 15 | 1914 | |||
| EF492985.1 | - | 15 | 1914 | |||