siRNA Details
| Gene Symbol: Tbkbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172021.2 | NP_742018.2 | 3 | 360 | ||
| XM_006247266.2 | XP_006247328.1 | 2 | 1510 | |||
| XM_008767953.1 | XP_008766175.1 | 2 | 1510 | |||
| GenBank mRNA | AJ278800.1 | - | 2 | 346 | ||
| BC076503.1 | - | 3 | 360 | 7100584 | ||

Tbkbp1
Genomic Map