siRNA Details
| Gene Symbol: Soat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153728.2 | NP_714950.2 | 4 | 574 | ||
| GenBank mRNA | AB075946.1 | - | 4 | 306 | ||
| AB101480.1 | - | 4 | 574 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Soat2| Gene Symbol: Soat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153728.2 | NP_714950.2 | 4 | 574 | ||
| GenBank mRNA | AB075946.1 | - | 4 | 306 | ||
| AB101480.1 | - | 4 | 574 | |||