siRNA Details
| Gene Symbol: Vom1r90 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153729.2 | NP_714951.1 | 3 | 1018 | ||
| GenBank mRNA | AY510354.1 | - | 1 | 790 | ||
| U36898.1 | - | 3 | 1038 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vom1r90| Gene Symbol: Vom1r90 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153729.2 | NP_714951.1 | 3 | 1018 | ||
| GenBank mRNA | AY510354.1 | - | 1 | 790 | ||
| U36898.1 | - | 3 | 1038 | |||