siRNA Details
Gene Symbol: Klhl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153730.2 | NP_714952.1 | 10 | 1420 | ||
XM_006249843.2 | XP_006249905.1 | 11 | 1569 | |||
XM_006249844.2 | XP_006249906.1 | 11 | 1843 | |||
XM_006249845.1 | XP_006249907.1 | 11 | 1549 | |||
XM_006249846.2 | XP_006249908.1 | 12 | 1635 | |||
XM_008769539.1 | XP_008767761.1 | 11 | 1608 | |||
XM_008769540.1 | XP_008767762.1 | 12 | 1463 | |||
XM_008769541.1 | XP_008767763.1 | 11 | 1353 | |||
GenBank mRNA | AJ133126.1 | - | 10 | 1462 | ||
BC086983.1 | - | 11 | 1550 | 7133597 | ||
FQ210179.1 | - | 12 | 1619 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
