siRNA Details
Gene Symbol: Klhl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153730.2 | NP_714952.1 | 7, 8 | 1147 | ||
XM_006249843.2 | XP_006249905.1 | 8, 9 | 1296 | |||
XM_006249844.2 | XP_006249906.1 | 8, 9 | 1570 | |||
XM_006249845.1 | XP_006249907.1 | 8, 9 | 1276 | |||
XM_006249846.2 | XP_006249908.1 | 9, 10 | 1362 | |||
XM_008769539.1 | XP_008767761.1 | 8, 9 | 1335 | |||
XM_008769540.1 | XP_008767762.1 | 9, 10 | 1190 | |||
XM_008769541.1 | XP_008767763.1 | 8, 9 | 1080 | |||
GenBank mRNA | AJ133126.1 | - | 7, 8 | 1189 | ||
BC086983.1 | - | 8, 9 | 1277 | 7133597 | ||
FQ210179.1 | - | 9, 10 | 1346 |
