siRNA Details
| Gene Symbol: Zfp597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153732.2 | NP_714954.2 | 3 | 619 | ||
| GenBank mRNA | AF277900.2 | - | 495 | |||
| BC078709.1 | - | 3 | 619 | 7113822 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp597| Gene Symbol: Zfp597 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153732.2 | NP_714954.2 | 3 | 619 | ||
| GenBank mRNA | AF277900.2 | - | 495 | |||
| BC078709.1 | - | 3 | 619 | 7113822 | ||