siRNA Details
| Gene Symbol: Serpinb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153733.2 | NP_714955.2 | 6 | 684 | ||
| GenBank mRNA | AY075037.1 | - | 6 | 700 | ||
| BC061735.1 | - | 6 | 684 | 5598648 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Serpinb10| Gene Symbol: Serpinb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153733.2 | NP_714955.2 | 6 | 684 | ||
| GenBank mRNA | AY075037.1 | - | 6 | 700 | ||
| BC061735.1 | - | 6 | 684 | 5598648 | ||