siRNA Details
| Gene Symbol: Rimbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100488.2 | NP_001093958.1 | 18 | 3432 | ||
| XM_006249257.2 | XP_006249319.1 | 18 | 4105 | |||
| XM_006249258.2 | XP_006249320.1 | 18 | 4288 | |||
| XM_008769189.1 | XP_008767411.1 | 18 | 4037 | |||
| XM_008769190.1 | XP_008767412.1 | 18 | 5205 | |||
| XM_008769191.1 | XP_008767413.1 | 18 | 4105 | |||
| XM_008769192.1 | XP_008767414.1 | 17 | 3394 | |||
| XM_008769193.1 | XP_008767415.1 | 17 | 3394 | |||
| GenBank mRNA | AF199336.1 | - | 3 | 376 | ||

Rimbp2
Genomic Map