siRNA Details
| Gene Symbol: Prl3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153736.1 | NP_714958.1 | 4 | 598 | ||
| GenBank mRNA | AB019791.1 | - | 4 | 600 | ||
| AF526270.1 | - | 4 | 598 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prl3a1| Gene Symbol: Prl3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153736.1 | NP_714958.1 | 4 | 598 | ||
| GenBank mRNA | AB019791.1 | - | 4 | 600 | ||
| AF526270.1 | - | 4 | 598 | |||