siRNA Details
| Gene Symbol: Eaf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172047.1 | NP_742044.1 | 4 | 546 | ||
| XM_008768707.1 | XP_008766929.1 | 4 | 433 | |||
| XM_008768708.1 | XP_008766930.1 | 2 | 1102 | |||
| XM_008768709.1 | XP_008766931.1 | 2 | 832 | |||
| GenBank mRNA | AY049022.1 | - | 4 | 546 | ||

Eaf2
Genomic Map