siRNA Details
| Gene Symbol: Zfp187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771777.1 | XP_008769999.1 | 7 | 1174 | ||
| XM_577579.6 | XP_577579.4 | 7 | 1008 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp187| Gene Symbol: Zfp187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771777.1 | XP_008769999.1 | 7 | 1174 | ||
| XM_577579.6 | XP_577579.4 | 7 | 1008 | |||