siRNA Details
| Gene Symbol: Alk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169101.2 | NP_001162572.1 | 3298 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Alk| Gene Symbol: Alk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001169101.2 | NP_001162572.1 | 3298 | |||