siRNA Details
| Gene Symbol: Sostdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153737.1 | NP_714959.1 | 1 | 221 | ||
| GenBank mRNA | AF411056.1 | - | 1 | 221 | ||
| BC081710.1 | - | 1 | 195 | 7191839 | ||
| FQ213516.1 | - | 1 | 211 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sostdc1| Gene Symbol: Sostdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153737.1 | NP_714959.1 | 1 | 221 | ||
| GenBank mRNA | AF411056.1 | - | 1 | 221 | ||
| BC081710.1 | - | 1 | 195 | 7191839 | ||
| FQ213516.1 | - | 1 | 211 | |||