siRNA Details
| Gene Symbol: Prl7b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153738.1 | NP_714960.1 | 6 | 623 | ||
| GenBank mRNA | AF525159.1 | - | 6 | 623 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prl7b1| Gene Symbol: Prl7b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153738.1 | NP_714960.1 | 6 | 623 | ||
| GenBank mRNA | AF525159.1 | - | 6 | 623 | ||