siRNA Details
| Gene Symbol: Sars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007606.1 | NP_001007607.1 | 6, 7 | 799 | ||
| XM_006233136.2 | XP_006233198.1 | 6, 7 | 834 | |||
| GenBank mRNA | AY145052.1 | - | 2, 3 | 226 | ||
| BC061765.1 | - | 6, 7 | 799 | 5620893 | ||

Sars
Genomic Map