siRNA Details
| Gene Symbol: Rims4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170666.1 | NP_733766.1 | 5 | 485 | ||
| XM_008762471.1 | XP_008760693.1 | 5 | 602 | |||
| GenBank mRNA | AF548739.1 | - | 5 | 485 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rims4| Gene Symbol: Rims4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170666.1 | NP_733766.1 | 5 | 485 | ||
| XM_008762471.1 | XP_008760693.1 | 5 | 602 | |||
| GenBank mRNA | AF548739.1 | - | 5 | 485 | ||