siRNA Details
| Gene Symbol: Kcnj14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170718.1 | NP_733836.1 | 2 | 1590 | ||
| GenBank mRNA | AJ003065.1 | - | 2 | 1590 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnj14| Gene Symbol: Kcnj14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170718.1 | NP_733836.1 | 2 | 1590 | ||
| GenBank mRNA | AJ003065.1 | - | 2 | 1590 | ||