siRNA Details
| Gene Symbol: Lzts3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172022.2 | NP_742019.2 | 2 | 101 | ||
| GenBank mRNA | AJ278801.2 | - | 2 | 101 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lzts3| Gene Symbol: Lzts3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172022.2 | NP_742019.2 | 2 | 101 | ||
| GenBank mRNA | AJ278801.2 | - | 2 | 101 | ||