siRNA Details
| Gene Symbol: Stab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001246357.1 | NP_001233286.1 | 3422 | |||
| GenBank mRNA | AY007370.1 | - | 1 | 56 | ||
| GQ477370.2 | - | 3422 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stab2| Gene Symbol: Stab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001246357.1 | NP_001233286.1 | 3422 | |||
| GenBank mRNA | AY007370.1 | - | 1 | 56 | ||
| GQ477370.2 | - | 3422 | ||||