siRNA Details
| Gene Symbol: Mtmr9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005761.1 | NP_001005761.1 | 4 | 684 | ||
| GenBank mRNA | AF526269.1 | - | 4 | 296 | ||
| BC083644.1 | - | 4 | 684 | 7134963 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mtmr9| Gene Symbol: Mtmr9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005761.1 | NP_001005761.1 | 4 | 684 | ||
| GenBank mRNA | AF526269.1 | - | 4 | 296 | ||
| BC083644.1 | - | 4 | 684 | 7134963 | ||