siRNA Details
Gene Symbol: Muc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760036.1 | XP_008758258.1 | 23 | 3592 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Muc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760036.1 | XP_008758258.1 | 23 | 3592 |