siRNA Details
| Gene Symbol: Muc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760036.1 | XP_008758258.1 | 8, 9 | 1689 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Muc6| Gene Symbol: Muc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760036.1 | XP_008758258.1 | 8, 9 | 1689 | ||