siRNA Details
| Gene Symbol: Cttnbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114401.1 | NP_001107873.1 | 4 | 788 | ||
| GenBank mRNA | AF053768.1 | - | 4 | 698 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cttnbp2| Gene Symbol: Cttnbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114401.1 | NP_001107873.1 | 4 | 788 | ||
| GenBank mRNA | AF053768.1 | - | 4 | 698 | ||