siRNA Details
| Gene Symbol: Impa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172224.1 | NP_757378.1 | 4 | 554 | ||
| XM_006254856.2 | XP_006254918.1 | 4 | 3873 | |||
| GenBank mRNA | AY160191.1 | - | 4 | 554 | ||
| BC083544.1 | - | 4 | 528 | 7191219 | ||

Impa2
Genomic Map