siRNA Details
| Gene Symbol: Impa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172224.1 | NP_757378.1 | 1, 2 | 287 | ||
| GenBank mRNA | AY160191.1 | - | 1, 2 | 287 | ||
| BC083544.1 | - | 1, 2 | 261 | 7191219 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Impa2| Gene Symbol: Impa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172224.1 | NP_757378.1 | 1, 2 | 287 | ||
| GenBank mRNA | AY160191.1 | - | 1, 2 | 287 | ||
| BC083544.1 | - | 1, 2 | 261 | 7191219 | ||